Would you like to log in again?
BoneClones® Comparative Skulls
Supplier: BONE CLONES, INC.
BoneClones®
COMP-160
470352-912KT 979.75 USD InStock
470352-912
BoneClones® Comparative Skulls
Models Zoology Models
Compare skulls and dentition from different animal types.
  • Five full size skulls
  • Reptile, avian, and mammal skulls
  • Mix of dietary dentition
Easily illustrate the differences in skull anatomy and dentition to students with one set of skulls. A mix of reptile (Alligator and Snapping Turtle), Avian (Blue Jay), herbivorous rodents (Jack Rabbit and Beaver), Omnivore mammal (Skunk), and Carnivorous Mammal (Gray Fox). Great as stations to identify unique structures, or in sets to show commonality.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty