Would you like to log in again?
Biochrom Cuvettes for Photometers and Spectrophotometers
Supplier: Biochrom
80-3000-83 80-3000-77 80-2084-11 80-2004-53 80-2002-95 80-2004-15
490005-912EA 2052.95 USD InStock
490005-912 490014-202 490005-884 490005-886 490014-204 490014-208
Biochrom Cuvettes for Photometers and Spectrophotometers
Cuvettes Spectrophotometer Cuvettes
Quartz glass cuvettes are suitable for use in the wavelength range 230 - 2500 nm. Optical glass cuvettes are suitable for use in the wavelength range 320 - 2500 nm.
  • Sizes range from 1.4 ml to 750 µl volume
Biochrom has a wide range of cuvettes for numerous spectrophotometers and photometers. Ranging from quartz, plastic, optical glass or acrylic, and various volume sizes, you are sure to find what you need from this selection of cuvettes!
Ordering information: Quartz (490005-884, 490005-912), plastic (490014-202, 490005-866), optical glass (490014-208), acrylic (490014-204). Pack of 100 (490005-886, 490014-202, 490014-204). 10 mm (490014-202). semi-micro (490014-202, 490005-886) or ultra micro (490005-912).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty