Would you like to log in again?
MicroPoint, extended length, ART 10 REACH, Thermo Scientific
Supplier: Thermo Scientific
ART®
3510 2140-05
72830-420CS 1255.21 USD InStock
72830-420 490014-542
MicroPoint, extended length, ART 10 REACH, Thermo Scientific
Pipette Tips
Molecular BioProducts ART (Aerosol Resistant Tip) specialty pipette tips seal when exposed to potential contaminants, trapping them inside the barrier, providing accurate and consistent performance.
These tips come with a self-sealing filter tip that protects pipettes from crossover contamination by not allowing liquids or aerosols to touch pipettes.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty