Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Alera® Rolling File Cart

Supplier: Alera
Alera®
ALEFW601426BL ALEFW601424BL
500010-621EA 75.6 USD InStock
500010-621 500010-625
Alera® Rolling File Cart
Carts
Mobile files help keep paper work handy when needed it and out of the way when you don't.

  • Four 2" hooded casters, two locking
  • Assembles without tools

Bottom shelf adjusts in 1" increments and are rated at 250 lbs

File size format is legal, letter

Certifications: GREENGUARD Children & Schools Certified, Meets or exceeds ANSI/BIFMA standards.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty