Would you like to log in again?
Parafilm® M Sealing Film
Supplier: Heathrow Scientific
Parafilm®
HS234526C HS120265 HS234526B HS234526A
52858-000CS 1323.54 USD InStock
52858-000 52858-032 52858-076 52859-079
Parafilm® M Sealing Film
Seals General Purpose Sealing Films
Parafilm® M is a semi-transparent, flexible, thermoplastic film with two-way stretch.
  • Creates tight moisture-barrier that holds a strong seal and doesn't release from glassware, despite off-gassing
  • Reduces the risk of tampered data with a covering that is odorless, colorless and semi-transparent
  • Prevents moisture and volume loss due to material composition and tight seal
  • Flexible and self-sealing material allows it to cling around irregular shapes and surfaces
  • Stretches more than 200% of original length
Non-toxic and free from plasticizers, primarily composed of polyolefins and paraffin waxes offering good resistance against acids, alkalis and some organic solvents - use not recommended with acetone, diethyl ether or tetrachloride. Resistant up to 48 hours against many polar substances such as saline solutions, inorganic acids and alkaline solutions.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty