Would you like to log in again?
Pure™ 200 Pipette Tips, Molecular BioProducts
Supplier: Molecular BioProducts
Pure™
3552 3550 3551
53225-690CS 1090.16 USD InStock
53225-690 53225-692 53225-694
Pure™ 200 Pipette Tips, Molecular BioProducts
Pipette Tips
These tips feature the MicroPoint* design and a 10 µl reference mark for accurate sample handling and precise delivery.
  • Sterile tips are certified RNase-, DNase-, DNA-, and pyrogen-free
  • Nonsterile tips are autoclavable
Ideal for a wide variety of applications. Tips fit Pipetman*, Eppendorf*, Finnpipette*, Nichiryo*, Rainin*, and Transferpette* pipettors.
Ordering information: Tips 53225-694 and -692 are packaged 96 per tray, 10 trays per pack. Tips 53225-690 are bulk packaged.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty