Would you like to log in again?
Pure™ 300E Pipette Tips, Molecular BioProducts
Supplier: Molecular BioProducts
Pure™
3562
53225-698CS 830.75 USD InStock
53225-698
Pure™ 300E Pipette Tips, Molecular BioProducts
Pipette Tips
The Pure 300E tips are ideal for multichannel pipettors.
  • Nonsterile
  • Autoclavable
They provide a secure fit and extra volume for repeat pipetting. Compatible with Transferpette*, Biohit*, Rainin* and others.
Ordering information: Tips are packaged 96 per tray, 10 trays per pack.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty