Would you like to log in again?
ART® 1000E Self-Sealing Barrier Pipette Tips, Molecular BioProducts
Supplier: Thermo Scientific ART
ART®
2079E 2079EIW
72830-042CS 1170.24 USD InStock
72830-042 53225-782
ART® 1000E Self-Sealing Barrier Pipette Tips, Molecular BioProducts
Pipette Tips
Tips ensure 100% protection against aerosol and liquid contamination.
  • Pre-sterilized tips
  • Tips provide an ideal fit for most 1000 µl pipettors
The ART® barrier traps contaminants, keeping them out of pipettors and samples. This also protects the sensitive inner components of the pipettor, prolonging pipettor life. ART® tips are ideal for nucleic amplification procedures, pipetting of radioactive samples, or any application where critical sample handling is required.
Certifications: Certified RNase-, DNase-, DNA-, and pyrogen-free.
Packaging: Sterile tips are packaged 100/tray, 8 trays per pack. Non sterile tips are bulk packed. The case of 500 tips are individually wrapped.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty