Would you like to log in again?
Accessories and Replacement Parts for VWR® Microdispensers
Supplier: VWR
3-033-388-G 3-033-590-G 3-033-275-G 3-033-320-G 3-00-010-590 3-033-250-G 3-033-210-G 3-033-225-G 3-033-105-G 3-033-333-G
53508-499PK 46.03 USD InStock
53508-499 53508-466 53508-400 53508-433 53508-660 53508-375 53508-386 53508-590 53498-162 53508-535
Accessories and Replacement Parts for VWR® Microdispensers
Pipettes Micropipettes
VWR® pipette tips and capillaries are for use with VWR® microdispensers (see 53506-100 series).
  • For use with VWR® microdispensers
Glass capillaries are color-coded, except for the 1000 µl size, which is made of natural polypropylene. The 20 µl capillary can only be used with VWR® 20 µl microdispenser (53506-530).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty