Would you like to log in again?
CENTRIFUGE EPP5810 220V/50HZ
Supplier: Eppendorf
Eppendorf* Multipurpose Centrifuge^ Model 5810. Capacity: 4 x400mL. Maximum Speed: 14,000rpm. Maximum RCF: 20,800xg. Speed Control: Variable, microprocessor controlled. Timer: 1 to 99min. and hold. 10 acceleration/braking rates. User defined programs. 220V/50Hz/450W/15A.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CENTRIFUGE EPP5810 220V/50HZ 53513-810 022625055 Each Retrieving
Green Product
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty