Would you like to log in again?
Open-Cell Antistatic Wafer Foams, Malaster Company, Inc.
Supplier: Malaster Company
MC5AFD MC6AFD MC605502ASL MC605602ASL
54200-100PK 25.6 USD InStock
54200-100 54200-102 54200-112 54200-108
Open-Cell Antistatic Wafer Foams, Malaster Company, Inc.
Wafer Handling Supplies Wafer Separators
Cost effective solution to secure storage and transport of semiconductor wafers.
  • Wafer foam cushion
  • Clean moisture barrier and shielding bags
  • Wafer separator
  • Shipping boxes
Highest level of protection against damage when used together with the wafer jar, foam cushions and separators. By using the wafer system, higher yields are achieved through a significant reduction in wafer breakage during storage, handling and transport between front end and back end sites.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty