Would you like to log in again?
Tyvek® Wafer Separators, Malaster Company
Supplier: Malaster Company
Tyvek®
MC6TD MC5TD
54200-110PK 55.63 USD InStock
54200-110 54200-106
Tyvek® Wafer Separators, Malaster Company
Wafer Handling Supplies Wafer Separators
  • Anti-static treated
  • 1056D Grade
  • Thickness: 6.3 mils
  • White color
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty