Would you like to log in again?
Nunc® MicroWell™ 96-Well Plates, Thermo Scientific
Supplier: Thermo Scientific
Nunc® MicroWell™
442587 266120 249570 268152 260844 260836 237108 249570 260895 262146 262146 269620 268152 260836 260844 268152 269620 260895 266120 442587 237108
73520-518CS 1011.72 USD InStock
CA62409-108 73520-518 CA73520-506 62409-108 73520-506 CA62409-114 62409-114 CA62409-068 62407-180 62409-068 CA62407-182 CA62407-180 62407-182 62407-166 82031-368 CA82031-368 CA62407-166 CATX73520-506 CA73520-486 73520-486 CA43300-451
Nunc® MicroWell™ 96-Well Plates, Thermo Scientific
Microplates HTS Microplates
These nonsterile 96-well plates are ideal for assays, colorimetric serology, and bacteriology applications
All plates are made of high-quality polystyrene to ensure optimum optical clarity, with the exception of plate 62409-108, which is made of polypropylene. Plate 260844 is generic bar coded.
Ordering information: Plate 62407-166 is provided with a lid. For lids and sealing tapes, see the 62409-118 series.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty