Would you like to log in again?
Nunc® MicroWell™ 96-Well Plates, Sterile, Thermo Scientific
Supplier: Thermo Scientific
Nunc® MicroWell™
260860 268200 262162 269787 243656 269787 260860 243656 249662 262162 268200
62407-174CS 933.1 USD InStock
CA62407-174 CA62407-184 62407-174 CA62409-116 62409-116 CA62409-111 62409-112 62409-111 CA62409-120 62409-120 62407-184
Nunc® MicroWell™ 96-Well Plates, Sterile, Thermo Scientific
Microplates HTS Microplates
These sterile, polystyrene plates are ideal for bacteriology, serology, molecular biology, and other laboratory applications
Plates have a standard format, which makes them compatible with plate washers and readers. V-bottom and U-bottom plates allow pelleting or settling of cells for gentle concentration and sample retrieval. Flat-bottom wells can be easily examined microscopically.
The wells have raised rims to reduce risk of cross-contamination. Lids have rings to control condensation during incubation. The beveled corners on the plates and lids prevent accidental reversal of the lid onto the plate. Wells are alphanumerically-labeled for easy identification. They stack conveniently and securely, with or without lids. Clone selection and replication can be performed using these plates with a few simple movements using a transferable solid phase (TSP) replicator.
Used for dilutions and general storage.
Ordering information: For lids and sealing tapes, see the Nunc® Microplate Lids, Thermo Scientific series.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty