Would you like to log in again?
Target2™ Syringe Filters, Cellulose Acetate
Supplier: National Scientific
Target2™
F2500-16 F2504-16 F2504-15 F2500-15
66064-851CS 2488.91 USD InStock
66064-851 66064-854 66064-834 66064-836
Target2™ Syringe Filters, Cellulose Acetate
Filters Syringe Filters
Cellulose acetate membrane for biological or protein-based samples.
  • Low nonspecific binding with limited chemical resistance
  • Good flow rate characteristics
Use the 4 mm filters for limited volume samples up to 2 ml, and the 13 and 25 mm filters for volumes up to 100 ml. The 0.2 µm filters can be used for 'cold' sterilization.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty