Would you like to log in again?
INDL INTERCHGE SOCKET 1/4INFPT
Supplier: 3M OCCUPATIONAL HEALTH
Industrial Interchange Socket^ This socket is a high pressure, 1/4 inch FPT brass fitting. This fitting is used on the 3M* Supplied Air Hose W-9435 (56621-987/980) and W-2929 (56621-994/90).
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
INDL INTERCHGE SOCKET 1/4INFPT 67101-342 7000056515 Case of 2 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty