Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2',7'-Dichlorofluorescein ACS

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: 2',7'-Dichloro-3',6'-dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one

DI116-500GMB DI116-25GM DI116-1GM DI116-5GM
700010-370EA 224.4 USD InStock
700010-370 700010-368 700010-366 700010-364
2',7'-Dichlorofluorescein ACS
2',7'-Dichlorofluorescein
2',7'-Dichlorofluorescein, Reagent, ACS, an organic dye of the fluorescein family, is used in the cellular antioxidant activity assay.

As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.


Formula: C₂₀H₁₀Cl₂O₅
MW: 401.20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005047
CAS Number: 76-54-0
Order Now
Specification Test Results
Clarity of Alcohol Solution To Pass Test
Suitability As Adsorption Indicator To Pass Test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty