Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-α-Terpineol ≥85%, Practical grade

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning
Synonyms: 2-(4-Methylcyclohex-3-en-1-yl)propan-2-olTerpineol

TE107-4LTGL TE107-500MLG TE107-55002 TE107-200LTB TE107-20LTMT
700016-050EA 179.52 USD InStock
700016-050 700016-046 700016-044 89092-200 700016-048
(±)-α-Terpineol ≥85%, Practical grade
(±)-α-Terpineol
alpha-Terpineol, Practical, is a naturally occurring monoterpene alcohol often isolated from pine oil. The Alpha isomer is one of three isomers, and is generally the most common constituent. It has a pleasenat lilac odor and for this reason is often used in perfumes, cosmetics and flavoring agents


Formula: C₁₀H₁₈O
MW: 154.25 g/mol
Density: 0,933
Storage Temperature: Ambient
MDL Number: MFCD00001557
CAS Number: 98-55-5
EINECS: 202-680-6
Order Now
Specification Test Results
Assay 85% Min.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty