Would you like to log in again?
S.F.P. Supplement (Perfringens S.F.P. Supplement), HiMedia
Supplier: HiMedia Laboratories
FD013-5VL FD013-5VL FD013-4X5VL FD013-4X5VL
71005-590PK 168.58 USD InStock
CA89128-828 71005-590 CA71005-590 89128-828
S.F.P. Supplement (Perfringens S.F.P. Supplement), HiMedia
Media for Microbiology Additives and Supplements for Microbiology
An antibiotic supplement, recommended for the selective isolation of C. perfringens.
  • Store at 2 to 8 °C
Caution: Not for medicinal Use.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty