Would you like to log in again?
BioRobotix® Pipette Tips for Tecan® and Cavro® Workstations, Molecular BioProducts
Supplier: Molecular BioProducts
BioRobotix®
904-021 902-021 902-251 904-261 904-251 904-253 901-021 901-251 901-252 903-021 903-261 903-252 902-011 903-251 903-011 903-253 902-253 901-253 901-011 904-252 902-252 904-011 901-262 901-261 902-261 904-262
72830-128CS 318.29 USD InStock
72830-128 72830-124 72830-146 72830-126 72830-148 72830-176 72830-132 72830-156 72830-178 72830-134 72830-172 72830-150 72830-174 72830-130 72830-158 72830-136 72830-142 72830-164 72830-144 72830-166 72830-182 72830-160 72830-162 72830-140 72830-184 72830-180
BioRobotix® Pipette Tips for Tecan® and Cavro® Workstations, Molecular BioProducts
Pipette Tips
Compatible with Tecan®, Cavro®, and Packard MultiPROBE II HT style workstations.
  • Sterile tips are certified RNase-, DNase-, DNA-, and pyrogen-free
Available as liquid sensing tips to maximize the workstation abilites, or clear polypropylene for applications that do not require liquid sensing. Total of six volumes available, for both tip styles, for increased repeatability in any application. Filter tips utilize the ART® barrier for increased protection against contamination.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty