Would you like to log in again?
Pipette Tips for Rainin LiteTouch™ Pipettors, Molecular BioProducts
Supplier: Molecular BioProducts
2779-05 3752 3781 2769 3782 3721 2749 2789 3751 2769-05 2779 2749-05
72830-276CS 1444.54 USD InStock
72830-276 72830-270 72830-282 72830-272 72830-471 72830-284 72830-280 72830-278 72830-454 72830-268 72830-444 82031-392
Pipette Tips for Rainin LiteTouch™ Pipettors, Molecular BioProducts
Pipette Tips
ART® self-sealing barriers trap contaminants, such as aerosols and liquids, keeping them out of pipettors and samples.
  • ART® tips are presterilized
  • Several tips are available with low retention technology
Tips are available in non sterile packaging, in sterile packaging, or with ART self-sealing barrier.
This allows the user to conduct PCR procedures and pipette radioactive or infectious samples. This permits more accurate sample dispensing, an increased level of reproducibility, and reduced sample loss due to increased adhesion to pipette tips.
Certifications: Certified RNase-, DNase-, DNA-, and pyrogen-free.
Packaging: All tips are packaged 96/rack.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty