Would you like to log in again?
Molecular BioProducts Low Retention Pipette Tips, Thermo Scientific
Supplier: Thermo Scientific
3512-05 3752-05 3751-05 3551-05 3771-05 3721-05 3510-05 3501-05 3500-05-11 3511-05
72830-430CS 569.54 USD InStock
72830-430 72830-460 72830-459 72830-426 72830-428 72830-432 72830-466 72830-422 72830-456 72830-446
Molecular BioProducts Low Retention Pipette Tips, Thermo Scientific
Pipette Tips
Molecular BioProducts low retention pipette tips are specifically designed for eliminating sample retention within the pipette tip.
All Pure tips are racked, pre-sterilized, and certified free of RNase, DNase, DNA, and pyrogen.
The inner surface of the tip is extremely hydrophobic, reducing loss of sample from adhesion to the tip interior. Tips are crystal clear to allow easy sample viewing.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty