Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VEGF (Human), ELISA Kit, Enzo Life Sciences

Supplier: Enzo Life Sciences
KIT156-0001
75786-352EA 638 USD InStock
75786-352
VEGF (Human), ELISA Kit, Enzo Life Sciences
Assays ELISAs
Ultra-sensitive ELISA kit for the quantitative detection of human VEGF with 100% cross reactivity to VEGF 165 and negligible cross-reactivity to VEGF 121.

  • Sensitive Assay: Detect VEGF as low as 4 pg /ml
  • Specific: 100% cross-reactivity to VEGF 165 with negligible cross-reactivity to related molecules including VEGF 121
  • Flexible: Wide dynamic range ideal for various matrices
  • Higher throughput: Assay up to 38 samples in duplicate with each kit with an assay time of just 2.5 hours
  • Efficiency: Color coded reagents help to reduce errors and increase efficiency

For the quantitative determination of human VEGF in serum, plasma, and culture supernatants.

The VEGF (human), EIA kit is a sensitive colorimetric immunometric enzyme immunoassay kit with results in just 2.5 hours. Absorbance is read at 450 nm. The wide dynamic range is ideal for various matrices. Assay up to 39 samples in duplicate with each kit.

Ordering information: Contains Microtiter Plate, Conjugate, Detector Antibody, Assay buffer, Antibody Diluent, Wash Buffer Concentrate, Standard, TMB Substrate, Stop Solution.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty