Would you like to log in again?
ADAPTER F/26X88MM 25ML TUBE EBA 280 CENT
Supplier: HETTICH INSTRUMENTS
Adapter 24 x 100 mm^ Adapter, Tube capacity: 25 ml, Tube Dimension: 24 x 100 mm, Maximum RCF: 3,703, For angle rotor, Mikro 220/220 R Microlitre Centrifuges
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
ADAPTER F/26X88MM 25ML TUBE EBA 280 CENT 75801-772 1633 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty