Would you like to log in again?
ADAPTER F/12.5X56MM 96TUB/RTR FOR BUCKET
Supplier: HETTICH INSTRUMENTS
Adapter 12x75mm/12x82mm/12x100mm/14x100mm^ Adapter, Tube capacity: 4,5,6,7ml, Tube Dimension: 12x75mm/12x82mm/12x100mm/14x100mm, Maximum RCF: 4,668, For swing out rotor, Rotina 380/380R Benchtop Centrifuges
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
ADAPTER F/12.5X56MM 96TUB/RTR FOR BUCKET 75801-830 1762-A Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty