Would you like to log in again?
ADAPTER F/45X87MM 100ML ROUND BTM TUBE
Supplier: HETTICH INSTRUMENTS
Adapter 44 x 100 mm^ Adapter, Tube capacity: 100 ml, Tube Dimension: 44 x 100 mm, Maximum RCF: 4,640, For swing out rotor, Rotina 380/380R Benchtop Centrifuges
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
ADAPTER F/45X87MM 100ML ROUND BTM TUBE 75801-842 1766 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty