Would you like to log in again?
ADAPTER F/50ML TUBE FALCON 30X87MM ROTOR
Supplier: HETTICH INSTRUMENTS
Adapter for Rotanta 460 rotor^ Adapter, for Rotanta 460 Swing out rotor, Maximum RCF: 5298, tube capacity: 50ml, Dimension: 29 x 115mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
ADAPTER F/50ML TUBE FALCON 30X87MM ROTOR 75802-014 4441 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty