Would you like to log in again?
ADAPTER F/ 250ML TUBE 62X122MM 4500 RPM
Supplier: HETTICH INSTRUMENTS
Adapter 61x122mm^ Adapter, bottle, Tube capacity: 250ml, Tube Dimensions: 61x122mm, Maximum RCF: 5,818, for Swing-out rotor, ROTO SILENTA 630 RS centrifuge
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
ADAPTER F/ 250ML TUBE 62X122MM 4500 RPM 75802-638 4238 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty