Super-Size AkroBins offer huge storage capacity and the versatility required to make the most of available space.
- Reinforced side ribs add strength and prevent spreading when stacked
- Large label area on front of bin
- Hinged lid covers and protects while allowing access to contents, even when bins are stacked
- Made in the USA
These heavy-duty plastic storage bins save space by securely stacking atop each other, and sitting on wire shelving and steel shelving, or steel carts and dollies. Super-Size AkroBins are resistant to weak acids and alkalis. Sturdy, one-piece construction is water, rust and corrosion-proof. Mobile options of the Super-Size AkroBins are also available. The mobile versions are equipped with 3" locking casters.
Accessories information: Keep items separated in bin using dividers, provides additional storage options.