Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 2PM ET on 20th April 2024.
VWR® Graduated Cylinders, Calibrated To Contain, Class B
Supplier: VWR
138.289.01A 138.289.04A 138.289.02A 138.289.07A 138.289.08A 138.289.05A VWRI75870-430 138.289.03A 138.289.06A
75870-430KT 162.42 USD InStock
75870-430 10545-960 10545-962 10545-952 10545-964 10545-954 10545-966 10545-956 10545-958
VWR® Graduated Cylinders, Calibrated To Contain, Class B
Cylinders Measuring Cylinders
Cylinder sizes from 25mL to 2000mL feature a hexagonal base and yellow bumper.
  • Borosilicate glass fabrication
  • Calibrated at 20°C to deliver
TC is printed on each cylinder which is ideal for receiving liquids where volumetric calculations are based solely on the volume contained within the cylinder
Made from ASTM E438 Type 1 Class A borosilicate 3.3 Glass. As per ASTM Specification E1272 Class B requirements.
Durable white color scale printing for long life in laboratory environment.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty