Would you like to log in again?
VWR® Low Form Griffin Beakers
Supplier: VWR
VWR®
1632417441950 1632411441020 1632411441010 1632417441400 VWRI75870-432 1632411441050 1632417441600 1632417441945 1632417441940 1632417441800 1632417441100 1632411441954 1632411441030 1632417441150 1632417441250
10754-954CS 279.63 USD InStock
10754-954 10754-700 10754-952 10754-948 10754-958 10754-946 10754-956 10754-760 10754-698 10754-962 10754-950 10754-696 10754-960 10754-762 75870-432
VWR® Low Form Griffin Beakers
Beakers
These low form Griffin beakers are made from high quality Type 1, Class A, 33 expansion borosilicate glass.
  • Double capacity scale
  • Uniform wall thickness
  • Very good chemical resistance
  • High temperature resistance
  • Meets ASTM E960, E960-93
They feature a thick, slightly flared, beaded top with a spout designed to have excellent pouring characteristics. They are printed with white graduated scale and a durable marking area.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty