Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 2PM ET on 20th April 2024.
VWR® Erlenmeyer Flasks, Narrow Mouth
Supplier: VWR
VWR®
231.402.01 231.435.07 VWRI75870-512 231.435.05 231.402.04A 231.402.02 231.402.03A 231.402.04 231.435.08A 231.402.09 231.435.06
10536-676CS 348.91 USD InStock
10536-676 10536-678 10536-924 10536-912 10536-926 10545-842 10536-914 10545-844 10545-846 10545-848 75870-512
VWR® Erlenmeyer Flasks, Narrow Mouth
Flasks Erlenmeyer Flasks
Flasks are durable and come with reinforced rim for easy pouring and also avoids any breakage while washing.
  • Borosilicate glass fabrication
  • Durable design
Feature durable white ceramic enamel scale which indicate approximate volume for measuring and mixing.
Manufactured from ASTM E438 Type 1 Class A borosilicate 33 expansion glass. As per ASTM specification E 1404 Type 1.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty