Would you like to log in again?
QuantiChrom™ Formaldehyde Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
DFOR-100
75877-970EA 517.38 USD InStock
75877-970
QuantiChrom™ Formaldehyde Assay Kit, BioAssay Systems
Assays Cellular Assays
Direct determination of formaldehyde concentrations in biological, food and environmental samples.
  • Safe, sensitive, non-radioactive assay
  • As low as 1.5 μM (45 ppb) formaldehyde can be quantified
  • "Mix-incubate-measure" type assay
  • No wash and reagent transfer steps are involved
  • Can be readily automated for processing thousands of samples per day
Formaldehyde (methanal) is the simplest aldehyde
BioAssay Systems' newly designed Formaldehyde Assay Kit provides a convenient fluorimetric means to measure formaldehyde in biological samples. In the assay, formaldehyde is derivatized with acetoacetanilide in the presence of ammonia. The resulting fluorescent product is then quantified fluorimetrically (&lambdaexc/em = 370/470nm). The assay is simple, sensitive, stable and high-throughput adaptable. The assay can detect as low as 1.5 μM formaldehyde in biological samples.
It is widely employed in industry for wide range of applications. Formaldehyde is also used as a disinfectant and is a commonly utilized tissue fixative and embalming agent. Formaldehyde is naturally present in all tissues and body fluids. Recently it has been shown that some cancer types exhibit elevated formaldehyde levels. Increased formaldehyde concentration in urine has been associated with prostate and bladder cancer. Thus, measuring formaldehyde in urine can be a very useful tool when studying cancer.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty