Would you like to log in again?
Phenylalanine Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
EPHE-100
75878-348EA 564.08 USD InStock
75878-348
Phenylalanine Assay Kit, BioAssay Systems
Assays Cellular Assays
determination of L-phenylalanine and evaluation of drug effects on its metabolism.
  • Safe. Non-radioactive assay.
  • Sensitive and accurate. Linear detection range of 2 - 300 μM L-phenylalanine.
  • Convenient and high-throughput. Homogeneous "mix-incubate-measure" type assay. No wash and reagent transfer steps are involved.
  • Can be readily automated on HTS liquid handling systems for processing thousands of samples per day.
  • Method: FL530/595nm
  • Samples: Serum, urine etc
L-Phenylalanine is one of the twenty common amino acids and an important precursor for several key signal molecules such as dopamine, norepinephrine, epinephrine, and the skin pigment melanin. It is found naturally in the breast milk of mammals, and used as nutritional supplements in food and drink products. The genetic disorder phenylketonuria is the inability to metabolize phenylalanine. Individuals with this disorder are known as "phenylketonurics". Individuals who cannot metabolize phenylalanine must monitor their intake of protein to control the buildup of phenylalanine. BioAssay Systems’ L-Phenylalanine Assay Kit provides a convenient fluorimetric means to measure L-phenylalanine in biological samples. In the assay, L-phenylalanine is oxidized by phenylalanine dehydrogenase, producing NADH, which reduces a fluorescent dye to a highly fluorescent product. The resulting fluorescence intensity (λex/em = 530/585 nm) is linear to the L-phenylalanine concentration in the sample.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty