Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ZMYND8 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-V98
75933-762EA 487 USD InStock
75933-762
Anti-ZMYND8 Rabbit Polyclonal Antibody
Antibodies
ZMYND8 is a receptor for activated protein kinase C (RACK). It also contains a bromodomain and two zinc fingers, and is thought to be a transcriptional regulator. Further, ZMYND8 is a cutaneous T-cell lymphoma-associated antigen. Anti-ZMYND8 Antibody is ideal for research in Gene Expression and Cell Signaling.

Anti-ZMYND8 Antibody is suitable for ELISA and Western Blots. Specific conditions for reactivity should be optimized by the end user.  Expect a band approximately 132 kDa in the appropriate cell lysate or extract.

Type: Primary
Antigen: ZMYND8
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty