Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD38 Mouse Monoclonal Antibody (Alexa Fluor® 546) [clone: HB7]

Supplier: Enzo Life Sciences
ABS158-0100
76002-448EA 658 USD InStock
76002-448
Anti-CD38 Mouse Monoclonal Antibody (Alexa Fluor® 546) [clone: HB7]
Antibodies
Recommended Applications: Flow Cytometry

Species reactivity: Human

Type: Primary
Antigen: CD38
Clonality: Monoclonal
Clone: HB7
Conjugation: Alexa Fluor® 546
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty