Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 2PM ET on 20th April 2024.
Nomex® Flame-Resistant Lab Coat, Men's/Unisex , CritiCore Protective Wear
Supplier: CritiCore Protective Wear
Nomex®
4940CXP63XL 4940CXP6L 4940CXP64XL 4940CXP6XL 4940CXP6XS 4940CXP6XXS 4940CXP6M 4940CXP62XL 4940CXP6S
76164-538CS 5529.94 USD InStock
76164-538 76164-544 76164-534 76164-546 76164-536 76164-540 76164-530 76164-542 76164-532
Nomex® Flame-Resistant Lab Coat, Men's/Unisex , CritiCore Protective Wear
Workwear
Designed to provide a superior level of protection against flash fire, electric arc (HRC 1), and other dangerous conditions, this Flame-Resistant (FR) lab coat has numerous applications in both laboratory and production settings. Made with Nomex® CXP6 fabric, it combines exceptional flame resistance with excellent durability, even after repeated industrial launderings.
  • Excellent heat-resistance properties
  • Best in class flammability protection
  • 360 degree FR identification and protection
  • Meets NFPA 70E and HRC 1 performance standards
  • Quick-release snap front
  • Safety sleeve cuff with tunneled elastic
Coats feature 360 degree FR identification with patches sewn on the back, plus left and right sleeves
Nomex® is a registered trademark owned by E. I. du Pont de Nemours and Company.
Constructed with a quick-release snap front and safety sleeve cuff with tunneled elastic for additional protection. Coats also feature two lower hip pockets and one breast pocket with pencil slot.
Certifications: Meets performance standards for NFPA 2112 and NFPA 70E HRC 1.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty