Would you like to log in again?
pH Probe with Flat Surface, Sper Scientific
Supplier: Sper Scientific
840051
76176-940EA 259.36 USD InStock
76176-940
pH Probe with Flat Surface, Sper Scientific
Electrodes pH Electrodes
pH probe is useful for reading the pH of small amounts of liquid, food products, and other surface pH measurements.
  • Easy to clean
  • Features a single-junction electrode for general-purpose applications
It features flat surface and is commonly used in testing surface measurements such as cheese, meats, small liquid samples, paper, soil, or skin.
Delivery information: Probe comes with a 40" cable and a standard BNC connector.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty