Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L(+)-Lysine monohydrochloride

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms: (S)-(+)-2,6-Diaminohexanoic acid monohydrochlorideH-Lys-OH.HClL-Lysine monohydrochlorideH-Lys-OH·HClL-Lysine monohydrocholoride(S)-(+)-Lysine monohydrochlorideLysine HydrochlorideL-Lysine Hydrochloride(S)-(+)-2,6-Diaminocaproic acid monohydrochloride

0219469791 0219469780 0219469790
76177-740EA 191.59 USD InStock
76177-740 76177-736 76177-738
L(+)-Lysine monohydrochloride
L(+)-Lysine monohydrochloride
Storage: Store at room temperature (15-30 °C)
L-Lysine monohydrochloride is widely used as nutritional supplements in food and beverage industries. It can also be used in animal feed as source of L-Lysine. L-Lysine Monohydrochloride can be used in a wide variety of industries including: food production, beverage, pharmaceutical, agriculture/animal feed, and various other industries.
L-Lysine monohydrochloride is a key amino acid in calcium absorption.

  • Cell Culture Reagent Grade
  • Presentation: White Powder
  • Soluble in aqueous solution (10 mg/mL-clear, colorless solution).


Formula: H₂N(CH₂)₄CH(NH₂)CO₂H·HCl
MW: 182.65 g/mol
Melting Pt: 263 °C (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00064564
CAS Number: 657-27-2
https://us-legacy.vwr.com/stibo/web/std.lang.all/08/61/36400861.jpg
Order Now
Specification Test Results
Identity Test Passes
Purity ≥99%
pH (10% aq soln) 5-6
Loss on Drying ≤1.0%
Optical Rotation +20° to +23° (c=8, 2N HCl)
Heavy Metals ≤15ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty