Synonyms:
(S)-(-)-2-Amino-4-(methylthio)butyric acid
, L(-)-Methionine
, Methionine
, (S)-(-)-2-Amino-4-(methylthio)butanoic acid
, H-Met-OH
, (S)-(-)-Methionine
, (S)-2-Amino-4-(methylthio)butanoic acid
0219470780
0219470725
0219470791
76177-872EA
623.73
USD
InStock
76177-872
76177-868
76177-914
L-Methionine ≥99%, white crystalline powder cell culture reagent
L-Methionine
Storage: Store at Room Temperature (15-30 °C).
Methionine is one of the common sulfur-containing amino acids. Methionine is a common methyl-group donor to various substrates, such as creatine, epinephrine, ergosterol, and choline.
- Cell Culture Reagent Grade
- Presentation: White Crystalline Powder
- pKa: 2.28, 9.21 (Lit.)
- Soluble at 2.5 g to 100 mL of water.