Would you like to log in again?
MICROMAN® Sterilized Capillary Pistons, Gilson®
Supplier: Gilson
Microman®
F148180 F148415 F148714 F148712 F148313 F148713 F148413 F148315
76178-168PK 181.07 USD InStock
76178-168 76178-156 76178-136 76178-158 76178-128 76178-164 76178-142 76178-130
MICROMAN® Sterilized Capillary Pistons, Gilson®
Pipette Tips
Capillary Pistons are designed, developed, and manufactured to fit the MICROMAN® perfectly.
Capillary pistons (CPs) provide 100% protection against contamination. With the disposable piston located inside the capillary tip, users can be assured that no harmful samples will contaminate the pipettor. The capillaries are made of pure polypropylene, are free of dyes, and fit even in the narrowest and deepest tubes.
They ensure optimal performance of the MICROMAN® pipettor across the full volume range.
Certifications: ISO 8655-2.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty