Would you like to log in again?
MICROMAN® Capillary Pistons, Gilson®
Supplier: Gilson
Microman®
F148560 F148012 F148014 F148412 F148113 F148414 F148114 F148312 F148013 F148314 F148112
76178-124PK 152.9 USD InStock
76178-124 76178-146 76178-134 76178-126 76178-138 76178-160 76178-162 76178-140 76178-166 76178-144 76178-132
MICROMAN® Capillary Pistons, Gilson®
Pipette Tips
Capillary Pistons are designed, developed, and manufactured to fit the MICROMAN® perfectly.
  • Made of pure PP
  • Slim fit allows you to use the narrowest and deepest test tubes
Designed to fit the Gilson MICROMAN positive displacement pipette, Capillary Piston Tips are ideal for difficult-to-pipette samples including; viscous, volatile, dense, hot or cold, or hazardous liquids. The tips work like a syringe providing protection against pipette, sample, and user contamination, and produce accurate, reproducible, and precise results.
Certifications: ISO 8655-2.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty