Would you like to log in again?
EXPERT™ Universal Fit Filter Tips, Gilson
Supplier: Gilson
Expert
F1731031 F1737032 F1731032 F1733031 F1735031 F1737031 F1733032 F1735032 F1735041 F1732032 F1735042 F1734031 F1732031 F1734032 F1736032 F1736031
76180-450PK 83.5 USD InStock
76180-450 76178-498 76178-376 76178-332 76178-500 76178-444 76178-378 76178-334 76178-328 76180-448 76178-492 76178-494 76178-372 76178-496 76178-374 76178-330
EXPERT™ Universal Fit Filter Tips, Gilson
Pipette Tips
EXPERT Filter Tips feature a patented, low-retention polymer that creates a hydrophobic surface to minimize sample binding when dispensing.
  • Economical and easy-to-use
  • Clear packaging allows you to easily identify the tips
  • Tips feature graduations at varying volumes allowing for quick-checks when aspirating samples
Sterilized EXPERT Tips are free of Human DNA, RNase, DNase, PCR inhibitors and pyrogens.
Certifications: Conforms with ISO 9001:2008.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty