Would you like to log in again?
PIPETMAN® Diamond Tips EasyPack™, Gilson®
Supplier: Gilson
PIPETMAN®
F161931 F161731 F161111 F161210 F161451 F161571 F161671 F161631
76178-148PK 72.76 USD InStock
76178-148 76180-024 76178-578 76180-020 76178-180 76178-182 76180-026 76178-152
PIPETMAN® Diamond Tips EasyPack™, Gilson®
Pipette Tips
Highest quality Diamond Tips for superior results.
  • Autoclaving conditions: Moist heat/121 °C/20 min/1 bar
  • EasyPack™ offers an easy way to order PIPETMAN® tips
Each box contains 1000 tips (5 bags of 200 tips/bag, or 1 bag of 1000 tips). Exceptions are the D10 ml tips, which contain 200 tips (4 bags of 50 tips/bag) and the D5000 tips (3 bags of 334 tips).
Bulk packs are sealed with a tamper-proof strip which can easily be opened for quick access to the tips. Bulk packs are closed using the rapid-sealing strip to prevent contamination and loss of tips.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty