Would you like to log in again?
SP Wilmad-LabGlass Benchtop Spectrometer NMR Tubes, SP Industries
Supplier: SP Industries
WG-BTNMR-8 WG-BTNMR-7
76183-196PK 302.84 USD InStock
76183-196 76183-194
SP Wilmad-LabGlass Benchtop Spectrometer NMR Tubes, SP Industries
Tubes NMR Tubes
Ideal for use with 43, 60, or 80 MHz manual sample loading benchtop NMR spectrometers, Wilmad-LabGlass Benchtop NMR tubes have been tested and approved by multiple benchtop spectrometer manufacturers, providing peace of mind and confidence when purchasing consumables for the instrument.
  • Economically priced for a high-throughput laboratory environment
  • 5 mm Ext.Ø tubes available in 7 or 8" lengths, in quantities of 25 or 150 pieces
  • Packaging allows for easy tube access and storage
  • Type 1, Class B Borosilicate glass construction with disposable caps
Caution: Not for use with high-field instruments or spinning experiments.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty