Would you like to log in again?
Type 29 Tiger Paw Angled Flap Discs, Weiler®
Supplier: Weiler
804-51104 804-51147 804-51148 804-51129 804-51118 804-51130 804-51149 804-51154 804-51133 804-51146 804-51123 804-51153 804-51145 804-51105 804-51150 804-51132
76183-844CT 88.62 USD InStock
76183-844 76183-878 76183-856 76183-868 76183-846 76183-858 76183-848 76183-880 76183-870 76183-872 76183-850 76183-874 76183-852 76183-952 76183-854 76183-876
Type 29 Tiger Paw Angled Flap Discs, Weiler®
Abrasives
100% zirconia alumina grain is fast cutting with aggressive action and long life, resulting in fewer changeovers and increased productivity
Use fo deburring, deflashing, edge chamfering, grinding and finishing in one step, stock removal, and weld blending applications on stainless steel.
Engineered poly/cotton cloth features excellent grain retention for maximum disc life.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty