Would you like to log in again?
Proto® Spline Sockets, Stanley® Products
Supplier: ORS Nasco
577-5020DPL 577-5530SPL 577-4710DPL 577-5420SPL 577-4708SPL 577-4708DPL 577-5014DPL 577-5524SPL 577-5732SPL 577-5528SPL 577-5018DPL 577-5324DPL 577-5534SPL 577-5526SPL 577-5426SPL 577-5012DPL
76188-986EA 61.93 USD InStock
76188-986 76188-984 76188-996 76189-578 76189-678 76189-426 76189-146 76189-674 76189-676 76190-746 76189-680 76189-670 76190-738 76189-562 76189-474 76189-672
Proto® Spline Sockets, Stanley® Products
Wrenches
Proto® Spline tools are designed to service high tensile strength fasteners used widely on military and commercial aircraft and in heavy machinery applications where vibration is prevalent.
  • Forged alloy steel
  • Inch measuring system
  • 12 points
Spline sockets help tighten high strength bolts to high torque loads. They are also used in automotive and industrial areas where high performance torque needs to be achieved. They help lower the force required to turn standard fasteners by allowing the torque load to be moved away from the corners to help prevent fastener rounding.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty