Would you like to log in again?
Proto® Electronic Fixed Ratchet Head Torque Wrenches, Essendant
Supplier: Stanley
577-H7-250FRB
76190-000EA 1165.63 USD InStock
76190-000
Proto® Electronic Fixed Ratchet Head Torque Wrenches, Essendant
Wrenches
Accuracy of +/- 2% of indicated value in both clockwise and counter clockwise directions. Broad accuracy range extends from 10% of full scale to 100% of full scale to help reduce the number of torque wrenches required. Ninety-nine "on-board" memory storage locations help improve efficiency when taking multiple torque readings. Three units of measure (Foot-lb., Inch-lb., and Nm) and three operating modes (track mode, peak hold, and auto clear). Red, green, and yellow lights along with an audible buzzer help identify when desired torque target zones are approached and met.
  • Fast sampling rate helps maintain high accuracy during torque applications
  • Ergonomically designed, comfortable cushion grip
  • Torque wrench head rotates to allow for easy viewing of LCD display
  • Target torque zone is adjustable from 1% to 10% of targeted torque value
  • Fixed ratcheting head with 24-tooth design and 15° arc swing
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty