Would you like to log in again?
Proto® Torqueplus™ Flex Sockets, ORS Nasco
Supplier: ORS Nasco
577-J4808AM 577-4778TA 577-5281A
76190-004EA 38.77 USD InStock
76190-004 76190-840 76190-702
Proto® Torqueplus™ Flex Sockets, ORS Nasco
Wrenches
Turning angle 65°.
  • Square drive
  • Nickel chrome finish
  • Box tip
  • Forged alloy steel
  • Ball locking type
Applies more torque to fastener. Concentric openings. High strength, thin wall design. Reduces risk of fastener rounding.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty