Would you like to log in again?
EARMUFF105 CAPMOUNT PELTOROPTIME BK 10CS
Supplier: 3M OCCUPATIONAL HEALTH
PELTOR* Optime* 105 Cap-Mount Earmuff^ PELTOR* Optime* 105 Cap-Mount Earmuff, For noise level up to 105dBA, No Dielectric, with Liquid/foam filled earmuff cushion, Color: Red/Black, Hearing Protection Style: Hard Hat Attached, Material: ABS, Noise Reduction Rating: 30 Decibel, Size: 1 Size Fits Most
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
EARMUFF105 CAPMOUNT PELTOROPTIME BK 10CS 76193-252 7000002327 Case of 10 Retrieving
Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty